Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Cnih3
Entrez Gene ID
690252
Gene Name
cornichon family AMPA receptor auxiliary protein 3
Gene Aliases
CNIH-3
Chromosome Location
Chr.13: 99663150 - 99770625 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001166578.1 NP_001160050.1 3 - 4 198 61
GenBank mRNA FJ403327.1 - 3 - 4 198 61

View Full Product Details