Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Hhipl2
Entrez Gene ID
317051
Gene Name
hedgehog interacting protein-like 2
Gene Aliases
RGD1311317
Chromosome Location
Chr.13: 101814574 - 101834641 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_229754.7 XP_229754.4 6 - 7 1716 64

View Full Product Details