Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Tp53bp2
Entrez Gene ID
305025
Gene Name
tumor protein p53 binding protein, 2
Gene Aliases
Trp53bp2
Chromosome Location
Chr.13: 100817204 - 100873837 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_223012.8 XP_223012.6 7 - 8 1070 66

View Full Product Details