The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001037204.1 | NP_001032281.1 | 9 - 10 | 1173 | 141 |
XM_008769862.1 | XP_008768084.1 | 9 - 10 | 1195 | 141 | |
GenBank mRNA | BC107466.1 | - | 9 - 10 | 1173 | 141 |