Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Pan3
Entrez Gene ID
360760
Gene Name
PAN3 poly(A) specific ribonuclease subunit
Gene Aliases
-
Chromosome Location
Chr.12: 9214112 - 9331904 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_006248842.2 XP_006248904.1 16 - 17 2678 118
XM_006248844.2 XP_006248906.1 16 - 17 2678 118
XM_346914.7 XP_346915.4 15 - 16 2515 118

View Full Product Details