Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Foxk1
Entrez Gene ID
304298
Gene Name
forkhead box K1
Gene Aliases
RGD1309643
Chromosome Location
Chr.12: 14183436 - 14244634 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001037219.2 NP_001032296.2 7 - 8 1675 125

View Full Product Details