The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_031144.3 | NP_112406.1 | 1 - 2 | 69 | 81 |
GenBank mRNA | BC063166.1 | - | 1 - 2 | 72 | 81 |
FQ231503.1 | - | 1 - 2 | 71 | 81 | |
FQ232650.1 | - | 1 - 2 | 71 | 81 |