Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Lrrc66
Entrez Gene ID
289587
Gene Name
leucine rich repeat containing 66
Gene Aliases
RGD1306094
Chromosome Location
Chr.14: 37128698 - 37153890 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001008763.1 NP_001008763.1 2 - 3 629 71
GenBank mRNA AY383699.1 - 2 - 3 629 71

View Full Product Details