Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Klf3
Entrez Gene ID
114845
Gene Name
Kruppel-like factor 3 (basic)
Gene Aliases
Bklf, Tef-2
Chromosome Location
Chr.14: 45142769 - 45173753 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001105742.1 NP_001099212.1 1 - 2 193 135

View Full Product Details