Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Wnt8b
Entrez Gene ID
293990
Gene Name
wingless-type MMTV integration site family, member 8B
Gene Aliases
-
Chromosome Location
Chr.1: 264244200 - 264264330 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001106359.1 NP_001099829.1 3 - 4 302 78
XM_006231442.2 XP_006231504.1 4 - 5 458 78

View Full Product Details