Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Amtn
Entrez Gene ID
689404
Gene Name
amelotin
Gene Aliases
-
Chromosome Location
Chr.14: 21286509 - 21300620 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001044296.1 NP_001037761.1 6 - 7 424 72
XM_008770057.1 XP_008768279.1 5 - 6 2080 72
GenBank mRNA AM231717.1 - 6 - 7 384 72
DQ198381.1 - 6 - 7 424 72

View Full Product Details