Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Enam
Entrez Gene ID
289525
Gene Name
enamelin
Gene Aliases
-
Chromosome Location
Chr.14: 21194635 - 21219529 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001106001.1 NP_001099471.1 9 - 10 882 73

View Full Product Details