Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Thap6
Entrez Gene ID
305244
Gene Name
THAP domain containing 6
Gene Aliases
-
Chromosome Location
Chr.14: 17595864 - 17616170 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001107209.2 NP_001100679.1 4 - 5 496 150
XM_008770033.1 XP_008768255.1 3 - 4 437 150
XM_008770035.1 XP_008768257.1 3 - 4 376 150
GenBank mRNA CK602350.1 - 3 - 4 395 150

View Full Product Details