Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Eif2b3
Entrez Gene ID
171145
Gene Name
eukaryotic translation initiation factor 2B, subunit 3
Gene Aliases
-
Chromosome Location
Chr.5: 135882955 - 135949276 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_133609.2 NP_598293.2 8 - 9 1081 71
XM_006238630.2 XP_006238692.1 8 - 9 1052 71
GenBank mRNA BC072507.1 - 8 - 9 1081 71
U38253.1 - 8 - 9 1061 71

View Full Product Details