Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Cldn19
Entrez Gene ID
298487
Gene Name
claudin 19
Gene Aliases
claudin-19
Chromosome Location
Chr.5: 138298630 - 138307982 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001008514.1 NP_001008514.1 2 - 3 472 59
XM_006238756.2 XP_006238818.1 2 - 3 944 59
XM_008763997.1 XP_008762219.1 3 - 4 929 59
GenBank mRNA AF497645.1 - 2 - 3 388 59
BC079172.1 - 2 - 3 472 59

View Full Product Details