The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001005543.1 | NP_001005543.1 | 12 - 13 | 2147 | 115 |
XM_006238646.2 | XP_006238708.1 | 12 - 13 | 2093 | 115 | |
XM_006238647.2 | XP_006238709.1 | 10 - 11 | 1129 | 115 | |
XM_008763993.1 | XP_008762215.1 | 12 - 13 | 2170 | 115 | |
GenBank mRNA | BC081913.1 | - | 12 - 13 | 2147 | 115 |
FQ212041.1 | - | 11 - 12 | 2087 | 115 |