Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Klhl5
Entrez Gene ID
305351
Gene Name
kelch-like family member 5
Gene Aliases
-
Chromosome Location
Chr.14: 44805213 - 44867078 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001047093.1 NP_001040558.1 2 - 3 728 91
XM_006250959.2 XP_006251021.1 2 - 3 384 91
XM_008770157.1 XP_008768379.1 2 - 3 206 91
GenBank mRNA AY641809.1 - 2 - 3 728 91

View Full Product Details