Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Pou2af1
Entrez Gene ID
690528
Gene Name
POU class 2 associating factor 1
Gene Aliases
-
Chromosome Location
Chr.8: 55603968 - 55631448 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001109599.1 NP_001103069.1 1 - 2 114 84

View Full Product Details