Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Ddx10
Entrez Gene ID
300710
Gene Name
DEAD (Asp-Glu-Ala-Asp) box polypeptide 10
Gene Aliases
-
Chromosome Location
Chr.8: 57667472 - 57830504 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001106820.1 NP_001100290.1 4 - 5 669 87

View Full Product Details