Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Zc3h12c
Entrez Gene ID
315658
Gene Name
zinc finger CCCH type containing 12C
Gene Aliases
RGD1565078
Chromosome Location
Chr.8: 56639047 - 56696968 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001108146.1 NP_001101616.1 3 - 4 976 83
XM_006243073.2 XP_006243135.1 3 - 4 1196 83

View Full Product Details