The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_052806.2 | NP_434693.1 | 5 - 6 | 1354 | 59 |
GenBank mRNA | AY574260.1 | - | 5 - 6 | 1354 | 59 |
U42976.1 | - | - | 1414 | 59 | |
X15834.1 | - | - | 1460 | 59 |