Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Aagab
Entrez Gene ID
171435
Gene Name
alpha- and gamma-adaptin binding protein
Gene Aliases
P34
Chromosome Location
Chr.8: 68526063 - 68564604 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_134398.2 NP_599225.1 8 - 9 855 103
XM_006243223.2 XP_006243285.1 8 - 9 884 103
GenBank mRNA AF178669.1 - 8 - 9 857 103
FQ224819.1 - 8 - 9 827 103

View Full Product Details