Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Crabp1
Entrez Gene ID
25061
Gene Name
cellular retinoic acid binding protein 1
Gene Aliases
-
Chromosome Location
Chr.8: 59344097 - 59352135 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001105716.1 NP_001099186.1 1 - 2 175 73

View Full Product Details