Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Las1l
Entrez Gene ID
296865
Gene Name
LAS1-like (S. cerevisiae)
Gene Aliases
RGD1565755
Chromosome Location
Chr.X: 65081591 - 65102461 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001170590.1 NP_001164061.1 2 - 3 387 76
XM_008773298.1 XP_008771520.1 2 - 3 378 76
GenBank mRNA BF554409.1 - - 76

View Full Product Details