Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Tmem38a
Entrez Gene ID
306327
Gene Name
transmembrane protein 38a
Gene Aliases
RGD1307901, Srp-27
Chromosome Location
Chr.16: 18960197 - 18974942 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001100175.1 NP_001093645.1 5 - 6 901 71
GenBank mRNA BC161801.1 - - 504 71
EF690436.1 - 5 - 6 901 71

View Full Product Details