Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Tcf7l1
Entrez Gene ID
312451
Gene Name
transcription factor 7-like 1 (T-cell specific, HMG-box)
Gene Aliases
Tcf3
Chromosome Location
Chr.4: 100492796 - 100660406 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001107865.1 NP_001101335.1 11 - 12 1339 70
XM_006236673.2 XP_006236735.1 12 - 13 1648 70
XM_006236674.2 XP_006236736.1 12 - 13 1645 70
XM_006236675.2 XP_006236737.1 11 - 12 1606 70

View Full Product Details