Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Gkn3
Entrez Gene ID
685128
Gene Name
gastrokine 3
Gene Aliases
Gkn3-ps1
Chromosome Location
Chr.4: 119181970 - 119187490 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_008763121.1 XP_008761343.1 3 - 4 409 75

View Full Product Details