Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Eif2ak3
Entrez Gene ID
29702
Gene Name
eukaryotic translation initiation factor 2 alpha kinase 3
Gene Aliases
PEK
Chromosome Location
Chr.4: 98648507 - 98709695 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_031599.2 NP_113787.1 5 - 6 1186 141
XM_006236623.1 XP_006236685.1 5 - 6 704 141
XM_006236624.2 XP_006236686.1 5 - 6 1224 141
XM_008762977.1 XP_008761199.1 4 - 5 818 141
GenBank mRNA AF096835.1 - 5 - 6 1187 141

View Full Product Details