The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001106819.1 | NP_001100289.1 | 2 - 3 | 553 | 73 |
XM_008766195.1 | XP_008764417.1 | 2 - 3 | 778 | 73 | |
GenBank mRNA | FQ218983.1 | - | 2 - 3 | 517 | 73 |