Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Tspan3
Entrez Gene ID
300733
Gene Name
tetraspanin 3
Gene Aliases
Tm4sf8
Chromosome Location
Chr.8: 60806848 - 60830448 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001005547.1 NP_001005547.1 5 - 6 685 106
GenBank mRNA BC082102.1 - 5 - 6 685 106

View Full Product Details