Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Hmbs
Entrez Gene ID
25709
Gene Name
hydroxymethylbilane synthase
Gene Aliases
PBG-D, PBGD, URO-S, hemC
Chromosome Location
Chr.8: 48667278 - 48674670 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_013168.2 NP_037300.2 5 - 6 395 89
XM_006242896.1 XP_006242958.1 5 - 6 481 89
XM_008766148.1 XP_008764370.1 4 - 5 565 89
XM_008766149.1 XP_008764371.1 4 - 5 502 89
GenBank mRNA BC088162.1 - 5 - 6 395 89
FQ231091.1 - 5 - 6 379 89
FQ234870.1 - 3 - 4 551 89
X06827.1 - 5 - 6 270 89
Y12006.1 - 5 - 6 265 89

View Full Product Details