Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Bace1
Entrez Gene ID
29392
Gene Name
beta-site APP cleaving enzyme 1
Gene Aliases
Bace
Chromosome Location
Chr.8: 50140092 - 50162388 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_019204.2 NP_062077.1 7 - 8 1516 68
GenBank mRNA AF190727.1 - 7 - 8 1516 68

View Full Product Details