Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Tbc1d21
Entrez Gene ID
363072
Gene Name
TBC1 domain family, member 21
Gene Aliases
RGD1310243
Chromosome Location
Chr.8: 63138801 - 63150694 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001014210.1 NP_001014232.1 - 144 103
GenBank mRNA BC078987.1 - - 144 103

View Full Product Details