The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001025424.1 | NP_001020595.1 | 4 - 5 | 484 | 146 |
GenBank mRNA | BC129075.1 | - | 4 - 5 | 458 | 146 |
DQ076481.1 | - | 4 - 5 | 484 | 146 |