Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Neil1
Entrez Gene ID
367090
Gene Name
nei endonuclease VIII-like 1 (E. coli)
Gene Aliases
-
Chromosome Location
Chr.8: 61817258 - 61824022 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001025754.1 NP_001020925.1 6 - 7 955 117
XM_006243134.2 XP_006243196.1 6 - 7 1769 117
XM_006243135.2 XP_006243197.1 6 - 7 1875 117
XM_006243136.2 XP_006243198.1 7 - 8 951 117
GenBank mRNA BC099125.1 - 6 - 7 955 117

View Full Product Details