The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001258011.1 | NP_001244940.1 | 4 - 5 | 606 | 127 |
GenBank mRNA | BC094214.1 | - | 4 - 5 | 606 | 127 |