Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
R3hdml
Entrez Gene ID
366245
Gene Name
R3H domain containing-like
Gene Aliases
-
Chromosome Location
Chr.3: 159886248 - 159897993 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001108962.1 NP_001102432.1 1 - 2 431 75
XM_006235600.2 XP_006235662.1 2 - 3 574 75

View Full Product Details