Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Wfdc3
Entrez Gene ID
296366
Gene Name
WAP four-disulfide core domain 3
Gene Aliases
-
Chromosome Location
Chr.3: 161193904 - 161211975 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001106541.1 NP_001100011.1 1 - 2 286 108
XM_006235533.2 XP_006235595.1 1 - 2 91 108

View Full Product Details