Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Ptgis
Entrez Gene ID
25527
Gene Name
prostaglandin I2 (prostacyclin) synthase
Gene Aliases
PGIS
Chromosome Location
Chr.3: 163950746 - 163986129 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_031557.2 NP_113745.1 2 - 3 460 62
GenBank mRNA BC061814.1 - 2 - 3 460 62
U53855.1 - 2 - 3 391 62

View Full Product Details