Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Grin2a
Entrez Gene ID
24409
Gene Name
glutamate receptor, ionotropic, N-methyl D-aspartate 2A
Gene Aliases
GluN2A, NMDAR2A, NR2A
Chromosome Location
Chr.10: 5707806 - 6119897 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_012573.3 NP_036705.3 2 - 3 636 94
GenBank mRNA D13211.1 - 1 - 2 429 94
M91561.1 - 2 - 3 636 94

View Full Product Details