Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Atf7ip2
Entrez Gene ID
497859
Gene Name
activating transcription factor 7 interacting protein 2
Gene Aliases
-
Chromosome Location
Chr.10: 5476205 - 5519938 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_006245728.2 XP_006245790.1 9 - 10 1086 151
XM_006245732.2 XP_006245794.1 9 - 10 1052 151
XM_006245735.1 XP_006245797.1 7 - 8 660 151
XM_008767496.1 XP_008765718.1 9 - 10 1085 151

View Full Product Details