Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Tekt5
Entrez Gene ID
363538
Gene Name
tektin 5
Gene Aliases
RGD1307081, Tek5, Tektin5
Chromosome Location
Chr.10: 5353007 - 5389684 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001014224.2 NP_001014246.1 3 - 4 714 108
GenBank mRNA AB294402.1 - 3 - 4 714 108
BC079040.1 - 3 - 4 786 108

View Full Product Details