Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Gng12
Entrez Gene ID
114120
Gene Name
guanine nucleotide binding protein (G protein), gamma 12
Gene Aliases
-
Chromosome Location
Chr.4: 97634925 - 97763478 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_008762971.1 XP_008761193.1 1 - 2 722 87

View Full Product Details