The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_024127.2 | NP_077041.2 | 2 - 3 | 292 | 100 |
GenBank mRNA | AB102787.1 | - | 2 - 3 | 142 | 100 |
AB102788.1 | - | 2 - 3 | 142 | 100 | |
BC081795.1 | - | 2 - 3 | 292 | 100 | |
L32591.1 | - | 2 - 3 | 160 | 100 |