Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Ankrd53
Entrez Gene ID
500230
Gene Name
ankyrin repeat domain 53
Gene Aliases
RGD1561200
Chromosome Location
Chr.4: 115439325 - 115447995 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
GenBank mRNA BC160898.1 - 5 - 6 1000 89

View Full Product Details