Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Arl8b
Entrez Gene ID
500282
Gene Name
ADP-ribosylation factor-like 8B
Gene Aliases
-
Chromosome Location
Chr.4: 140837745 - 140881866 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001024332.1 NP_001019503.1 1 - 2 309 129
GenBank mRNA BC081947.1 - 1 - 2 309 129
FQ220718.1 - 1 - 2 301 129

View Full Product Details