Assay Design
The assay probe spans an exon junction. This assay will detect gDNA.
Gene Transcripts
- Gene Symbol
- Gcs1
- Entrez Gene ID
-
78947
- Gene Name
- glucosidase 1
- Gene Aliases
-
Mogs
- Chromosome Location
-
Chr.4: 113948328 - 113951731 on Build Rnor_6.0
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
RefSeq |
NM_031749.2 |
NP_113937.1 |
3 - 4 |
778 |
92 |
GenBank mRNA |
AF087431.1 |
- |
3 - 4 |
779 |
92 |
- Gene Symbol
- LOC103692171
- Entrez Gene ID
-
103692171
- Gene Name
- mannosyl-oligosaccharide glucosidase
- Gene Aliases
-
-
- Chromosome Location
-
Chr.4: 114813988 - 114817463 on Build Rnor_6.0
View Full Product Details