The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | XM_006236871.2 | XP_006236933.1 | 1 - 2 | 118 | 91 |
XM_006236874.2 | XP_006236936.1 | 1 - 2 | 119 | 91 |