Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Slc41a3
Entrez Gene ID
641603
Gene Name
solute carrier family 41, member 3
Gene Aliases
-
Chromosome Location
Chr.4: 123467552 - 123513451 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001037492.1 NP_001032569.1 3 - 4 592 64
XM_006236871.2 XP_006236933.1 4 - 5 602 64
XM_006236872.2 XP_006236934.1 4 - 5 659 64
XM_006236873.1 XP_006236935.1 4 - 5 976 64
XM_006236875.2 XP_006236937.1 4 - 5 808 64
XM_006236876.2 XP_006236938.1 3 - 4 203 64
XM_006236877.2 XP_006236939.1 3 - 4 948 64
XM_006236878.2 XP_006236940.1 3 - 4 249 64
XM_006236879.2 XP_006236941.1 3 - 4 244 64
XM_008763134.1 XP_008761356.1 4 - 5 803 64
GenBank mRNA BC104698.1 - 3 - 4 592 64

View Full Product Details