Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Rab17
Entrez Gene ID
503269
Gene Name
RAB17, member RAS oncogene family
Gene Aliases
RGD1562478
Chromosome Location
Chr.9: 98118907 - 98131883 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001109363.2 NP_001102833.1 1 - 2 183 94
GenBank mRNA BC167022.1 - 1 - 2 183 94

View Full Product Details